Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Click to view available options
Volume (sistema metrico):
250 μL
300 μL
1000 μL
1100 μL
Filtrato:
Con filtro
Non-filtered
Imballaggio:
96 Tips/Tray, 10 Trays/Case
96 Tips/Tray, 50 Trays/Case
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Dimensione della confezione:
pezzi 4800
pezzi 960
Descrizione
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:
- Available as sterile and filtered
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifica
Specifica
| Volume (sistema metrico) | 300 μL |
| Filtrato | Non-filtered |
| Sterilità | Sterile |
| Colore | Trasparente |
| Formato | In rack |
| Stile punta | Automation |
| Imballaggio | 96 Tips/Tray, 10 Trays/Case |
| Da utilizzare con (apparecchiatura) | Universal Fit, Qiagen |
| Product Line | Qiagen |
| Fit for Purpose | Sterile |