Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Click to view available options
Volume (sistema metrico):
50 μL
80 μL
100 μL
150 μL
200 μL
200 µl
Sterilità:
Non sterile
Sterile
Sí
Filtrato:
Con filtro
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Dimensione della confezione:
pezzi 4800
Descrizione
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
- Available as standard, filtered and/or liquid sensing
- Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
- Lot-tested during production on actual workstation for which they are created
- Fully conductive, providing increased pipetting performance
- Low percentage coefficient of variation (%CV)
Specifica
Specifica
| Volume (sistema metrico) | 50 μL |
| Filtrato | Non-filtered |
| Sterilità | Sterile |
| Colore | Trasparente |
| Formato | In rack |
| Stile punta | Automation |
| Da utilizzare con (apparecchiatura) | Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius |
| Product Line | Tecan |
| Fit for Purpose | Sterile |