Learn More
Thermo Scientific™ Puntali per automazione per sistema di pipettaggio automatico Eppendorf™ epMotion™, 50, con filtro, sterili
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Descrizione
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
- Available in nonsterile, sterile and sterile filtered
- Validated on actual workstation for which tip is created
- Low percentage coefficient of variation
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifica
Specifica
| Volume (sistema metrico) | 1000 μL |
| Filtrato | Con filtro |
| Sterilità | Sterile |
| Colore | Trasparente |
| Formato | In rack |
| Stile punta | Automation |
| Da utilizzare con (apparecchiatura) | Eppendorf epMotion |
| Product Line | Eppendorf epMotion |
| Fit for Purpose | Sterile and PCR Ready |