missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Avian Influenza A Hemagglutinin Antibody Pack
For Research applications
Marca: Novus Biologicals™ NBP2-25066
Includes:
- 1.NB100-1933: Avian Flu Matrix Protein 1 Antibody
- 2.NB100-56572: Avian Flu Nucleoprotein Antibody
- 3.NB100-56607: Avian Flu HA protein Antibody
- 4.NB100-56608: Avian Flu HA protein Antibody
- 5.NB100-56610: Avian Flu Matrix Protein 1 Antibody
- 6.NB100-56611: Avian Flu Nucleoprotein Antibody
Descrizione
Specifica
| Avian Influenza A Hemagglutinin | |
| Unconjugated | |
| ha; HA1; HA2; Hemagglutinin; Hemagglutinin ha1 chain; Hemagglutinin ha2 chain; Hemagglutinin precursor | |
| 6 Vials | |
| Pig, Avian, Chicken |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
| Western Blot | |
| Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 72-88 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 128-143 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 9-28 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 212-225 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 58-77 Format: Purified Rabbit IgG polyclonal antibody; Application: ELISA, IF/ICC, IHC; Immunogen: amino acids 428-441 | |
| 1.NB100-1933: Avian Flu Matrix Protein 1 Antibody 2.NB100-56572: Avian Flu Nucleoprotein Antibody 3.NB100-56607: Avian Flu HA protein Antibody 4.NB100-56608: Avian Flu HA protein Antibody 5.NB100-56610: Avian Flu Matrix Protein 1 Antibody 6.NB100-56611: Avian Flu Nucleoprotein Antibody |
|
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto