Learn More
Carl Zeiss™ Axio Vert.A1 FL-LED Inverted Microscope
Codice prodotto. 12838065
Click to view available options
:
For Use With transmitted-light brightfield LED, phase contrast Ph1 and epi-fluorescence LED with intermediate phototube
Codice prodotto. 12838065
Marca: Carl Zeiss™ 4912370014000
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Compact inverted microscope ideal for routine research. This system is configured for transmitted light brightfield, phase contrast and epi-fluorescence (470 nm). The system has an intermediate tube included to allow the addition of a camera.
  • The Axio Vert.A1 is an inverted microscope ideal for advanced routine research and is the only system in its class with such a large range of features but still compact enough to sit directly beside your incubator. The system offers the ultimate flexibility and all standard contrast techniques including brightfield, phase contrast, PlasDIC, iHMC, DIC and epi-fluorescence are available.
  • The system has been designed with ergonomics and ease of use in mind. Whether you are sitting or standing, simply use intermediate pieces to work comfortably in an upright position. The ergo tube lets you tilt the eyepieces as much as you need. You can always view your samples easily. Thanks to ergonomic design of all operating parts, your routine tasks and work flows will be easier.
  • The Axio Vert.A1 offers either Halaogen or LED illumination for transmitted light. There are then two options for epi-fluorescence - either the Axio Vert.A1 FL-LED with inbuilt LEDs or the Axio Vert.A1 Fl with the ability to use an external light source for fluorescence to give more flexibility.
  • The accessories available with the Axio Vert.A1 are vast and these include multiple binocular tubes, documentation ports and attachments for infinity space. There are a range of options for stages with inserts available for a range of samples.
Specifica
Da utilizzare con (apparecchiatura) For transmitted-light brightfield LED, phase contrast Ph1 and epi-fluorescence LED with intermediate phototube