Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Sfoglia Tutto Axygen Scientific Prodotti
Click to view available options
Sterilità:
Non-sterile
Sterile
Colore chiusura:
Assorted
Blue
Brown
Clear
Gray
Green
Orange
Red
Violet
White
Yellow
Dimensione della confezione:
pezzi 4000
pezzi 500
Descrizione
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile tubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Wide range of colored tubes for easy sample identification
Specifica
Specifica
| Certificazione/Conformità | RNase, DNase, pyrogen-free |
| Colore chiusura | Gray |
| Materiale chiusura | Polypropylene |
| Capacità (sistema metrico) | 0.5 mL |
| Materiale | Polypropylene |
| Formato | Non-looped |
| Include | O-ring |
| Sterilità | Sterile |
| Tipo di chiusura | Screw Cap |
| Forma del fondo | Conical |
| Vedi altri risultati |