Learn More
Axygen™ Capless Tube for ABI Prism 310 Sequencer
Sfoglia Tutto Axygen Scientific Prodotti
Click to view available options
Capacità:
0.5 mL
Dimensione della confezione:
pezzi 2500
Descrizione
Axygen™ 0.5 mL polypropylene thin wall PCR tubes are designed to fit standard 96 well blocks.
RNase-/DNase-free Nonpyrogenic Nonsterile Manufactured with 99.9% pure virgin polypropylene Compatible with ABI-310 Instrument Caps for these tubes are AM-SEPTA-310 Axygen™ AxyMatsTM Single Septa Cap Compatible with ABI-310 Sequencer, Nonsterile
Specifica
Specifica
| Capacità | 0.5 mL |
| Da utilizzare con (apparecchiatura) | ABI Prism 310 sequencer |
| Product Type | Capillary Sequencing Tube |