Description
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook facilitates relaxed grip
- Large push button is easy on the thumb
- Locking feature prevents accidental volume changes
Specifications
Specifications
| Accuracy | ±1.2/±0.6% |
| Numero di canali | 1 |
| Tipo | Micropipette |
| Volume (sistema metrico) | Da 20 a 200 μl |
| Precisione | ±0.6/±0.2% |
| Quantità | 1 Pack |