Learn More
Axygen™ Axypet™ 8-Channel Digital Pipettor, Calibration 3x4 ALT
Codice prodotto. 17213634 Sfoglia Tutto Axygen Scientific Prodotti
Click to view available options
Volume (sistema metrico):
0.5 to 10 μL
5 to 50 μL
50 to 300 μL
Da 20 a 200 μl
Dimensione della confezione:
Pezzo
Codice prodotto. 17213634
Marca: Axygen™ AP810ALT
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
  • Push button and ejector button are ergonomically located
  • Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
  • Tip manifold rotates 360°
  • Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
  • Shafts withdraw when pressed against a row of tips
  • Each shaft moves independently, minimizing jamming of tips
  • Micrometer is continuously adjustable for selection of whole or fractional volumes
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità ISO17025
Numero di canali 8
Product Line Axypet
Tipo Multi-channel Pipettor
Volume (sistema metrico) 0.5 to 10 μL
Quantità 1/Pk., 1/Cs