Learn More
Axygen™ Axypet™ Starter Kit
Sfoglia Tutto Axygen Scientific Prodotti
Click to view available options
Volume (sistema metrico):
10 to 1000 μL
20 to 1000 μL
Dimensione della confezione:
1 set
Descrizione
- Micropipette starter kit
- Includes micropipettes, tips, calibration tools, identification tools, and more
Specifica
Specifica
| Product Line | Axypet |
| Tipo | Starter Kit, Micro |
| Volume (sistema metrico) | 20 to 1000 μL |
| Include | AP-20, AP-200, AP-1000 micropipettes, 1 rack each of 200μL and 1000μL tips, a calibration tool, ejector spacers, identification labels, lubricant, color identification rings, right-hand ejector button |
| Quantità | 1 Kit |