missing translation for 'onlineSavingsMsg'
Learn More

AZD-3463, MedChemExpress

Codice prodotto. 16422767
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16422767

Marca: Medchem Express HY15609

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

Specifica

Materiale o nome chimico AZD-3463
CAS 1356962-20-3
Colore Green
Note sul grado di purezza Research
Formula molecolare C24H25ClN6O
Quantità 50 mg
Sinonimo ALK/IGF1R inhibitor
Informazioni di solubilità DMSO : 20 mg/mL (44.55 mM; Need ultrasonic)
SMILES ClC(C=NC(NC1=C(OC)C=C(N2CCC(N)CC2)C=C1)=N3)=C3C4=CNC5=C4C=CC=C5
Molecular Weight (g/mol) 448.95
Peso formulazione 448.95
Percent Purity 98.27%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Cancer-Kinase/protease
Vedi altri risultati Mostra meno risultati

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.