missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan medoxomil, MedChemExpress

Codice prodotto. p-200097436
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16488747

Marca: Medchem Express HY14736

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Specifications

Materiale o nome chimico Azilsartan medoxomil
CAS 863031-21-4
Colore White
Note sul grado di purezza Research
Formula molecolare C30H24N4O8
Quantità 5 mg
Sinonimo TAK-491
Informazioni di solubilità DMSO : 125 mg/mL (219.87 mM; Need ultrasonic)
SMILES O=C(C1=C2C(N=C(OCC)N2CC3=CC=C(C4=CC=CC=C4C5=NC(ON5)=O)C=C3)=CC=C1)OCC6=C(C)OC(O6)=O
Molecular Weight (g/mol) 568.53
Peso formulazione 568.53
Percent Purity 99.88%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.