Learn More
Azlon™ Tappi in polietilene Azlon™
Click to view available options
Giunto rastremato standard:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Dimensione della confezione:
pezzi 10
Descrizione
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
- Manufactured from polyethylene
- Available in white and red
- Suitable for use with both Azlon and glass volumetric ware
- Enlarged head provides a good grip and protects glassware
- Ribbed cone ensures a secure leak free seal
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Matracci volumetrici e cilindri graduati in vetro e in plastica |
| Giunto rastremato standard | 34/35 |
| Colore | White, Red |
| Materiale | Polyethylene |
| Product Type | Tappo |
| Forma | Cono scanalato |