Learn More
Caraffe con manico in polipropilene con scala graduata incisa Azlon™
Click to view available options
Capacità (sistema metrico):
50 ml
100 ml
250 ml
500 ml
1000 ml
2000 ml
3000 ml
5000 ml
Graduazioni:
10mL
50mL
100mL
20 ml
500mL
500 ml
Dimensione della confezione:
Pezzo
pezzi 5
Descrizione
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
- Tapered jugs in high clarity polypropylene
- Permanent molded graduations
- Autoclavable at 121° C
- Ergonomically designed handle with thumb grip for easy carrying
Specifica
Specifica
| Capacità (sistema metrico) | 50 ml |
| Materiale | Polipropilene |
| Quantità | 5/conf. |
| Temperature Range (Metric) | Da – 20 a 135 °C |
| Graduazioni | 10mL |
| Graduated | Sì |
| Altezza (sistema metrico) | 68 mm |
| Diametro (sistema metrico) | 45 mm |
| Autoclavabile | Autoclavable |