Learn More
Dynalon™ Azlon™ Solvent Venting Wash Bottles
Sfoglia Tutto Cole-Parmer ProdottiDescrizione
Translucent low density polyethylene wash bottles feature unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. Wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.
Order Info
Bottles are available in 250 or 500mL capacities with five common solvent labels.
Compliance
All bottles have SPI recycling code stamped on base.
Specifica
Specifica
| Capacità | 8 once, 250 ml |
| Capacità (sistema metrico) | 250 mL |
| Capacità (sistema imperiale) | 8 once |
| Colore | Translucent |
| Materiale | LDPE |
| Larghezza (sistema metrico) | 57.15mm |
| Colore chiusura | Verde |
| Certificazione/Conformità | SPI recycling code stamped on base |
| Altezza (sistema metrico) | 203.2 mm |
| Larghezza (sistema imperiale) | 2.25 in. |
| Vedi altri risultati |