Learn More
Speirs Robertson™ Balance Platform
Codice prodotto. 11937047
Encompass_Preferred
Cambia vista
Click to view available options
Dimensions (L x W x H): 450 x 300 x 55mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
11937047 Dimensions (L x W x H): 450 x 300 x 55mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11937047 Fornitore Speirs Robertson™ N. del fornitore AMS30X45
Spedizione stimata: 16-12-2026
Ensures that vibrations common in most laboratories are effectively removed, greatly increasing instrument performance. Speirs Robertson™ Balance Platform is constructed of stainless steel with damped sorbothane for vibration reduction.
  • Unique Sorbothane isolator design removes up to 98 percent of unwanted vibrations
  • Custom Damped Sorbothane low frequency isolators built in
  • Pharma grade stainless meets stringent cleanliness requirements and is resistant to almost all forms of chemical attack
  • Sound deadened layer construction eliminates surface vibrations