Learn More
Thermo Scientific Chemicals Ball pestle; clear plexiglas; 1/8in. diameter
Click to view available options
Descrizione articolo:
100/Pk.
Quantity: 25/Pk.
Dimensione della confezione:
pezzi 100
pezzi 25
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Descrizione articolo | 100/Pk. |
| Diametro (sistema imperiale) esterno | 0.125 in. |
| Diametro (sistema metrico) esterno | 3.17mm |
| Product Type | Ball Pestle |
| Da utilizzare con (apparecchiatura) | Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT) |
Titolo del prodotto
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu