Learn More
Thermo Scientific™ Barcoded Latch Racks for Matrix™ Storage Tubes
GREENER_CHOICE
Codice prodotto. p-6524185
Click to view available options
Da utilizzare con (apparecchiatura):
0.75mL tubes
1.0mL Screwtop Tubes
1.4mL tubes
500μL Screwtop Tubes
500μL Tubes
N. per custodia:
5
10
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 5
Codice prodotto. 15543734
Marca: Thermo Scientific™ 4897BR
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.
Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.
Superior Storage Rack Design
  • Storage in robot-friendly ANSI-footprint rack
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
  • Rack barcodes are compatible with our range of VisionMate readers
  • Hinged lid rests in open position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench
Latch Racks Save Precious Space
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Latch rack lid can be lifted robotically for tube access with high-throughput automated applications
TRUSTED_SUSTAINABILITY
Specifica
Colore White
Descrizione Empty Latch Rack
Capacità (sistema metrico) 1 mL
Materiale Plastic
Da utilizzare con (apparecchiatura) 1.0mL Screwtop Tubes
N. per custodia 5
Codice a barre 2D Datamatrix; Linear EAN Code 128; Human Readable
Capacità 2D Matrix tubes
Linea di prodotti Matrix
Quantità confezione 5 Case
Capacità (sistema imperiale) 0.0338 fl oz.
Tipo di chiusura Screw Cap