Learn More
Emma Safety Footwear Bari (Frontier 118) Safety Shoes
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Thermoplastic Polyurethane Outsole
- ESD certified
- Chrome-free lining
- Sanitized
Specifica
Specifica
| Certificazione/Conformità | EN ISO 20345:2011, EN 12568:2010 |
| Tipo di chiusura | Laces |
| Colore | Blue |
| Progettazione | Bari (Frontier 118) |
| Materiale rivestimento interno | Hydro-Tec™ Sanitized Silver & Wellness leather |
| Materiale | Leather |
| Size | 41 |
| Materiale dello zoccolo | Steel |