Learn More
Barium fluoride crystal optic disc, 32mm x 3mm, polished both sides
Click to view available options
Quantità:
1 Ea.
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale o nome chimico | Barium fluoride crystal optic disc |
| Nota nome | 32mm x 3mm, Polished both sides |
| Quantità | 1 Ea. |