Learn More
Cytiva (formerly Pall Lab) Base without Support Screen
Replacement base with unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.
Marke: Cytiva (formerly Pall Lab) 4244
Weitere Details : Peso : 0.45600kg
Beschreibung
- Not approved for use in medical, clinical, surgical or other patient applications
Spezifikation
Polyphenylsulfone Stem | |
Non-sterile |