Learn More
Eppendorf™ Consegna di guanti bianchi per unità
Codice prodotto. p-7221200
Click to view available options
Service Offering:
White Glove Delivery (>180 kg)
White Glove Delivery (<100 kg)
White Glove Delivery (100 to 180 kg)
Dimensione della confezione:
Pezzo
Codice prodotto. 15773685
Marca: Eppendorf™ 2277000002
Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging. Shipment with stair climb to be ordered when needed.
  • White glove delivery per unit
  • Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging
  • Shipment with stair climb to be ordered when needed
TRUSTED_SUSTAINABILITY
Specifica
Service Offering White Glove Delivery (<100 kg)