Learn More
Nickel Electro™ Bath Armor
Sfoglia Tutto Nickel-Electro Prodotti
Click to view available options
Materiale:
Aluminum
Dimensione della confezione:
12L
Descrizione
- No more filling topping up water baths, maintaining water level
- Cleaning frequency reduced
- No water bourne contamination
- Good thermal heat distribution
- Replace water in your water bath with these thermally efficient pellets that are easy to keep clean
- It enables you to eliminate nearly all water bath cleaning and protects your water bath from drying out
- Saving the need for racks, floaties and bottle weights
- Aims to tackle water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
- The result is a dry bath that is far less conducive to contamination than a water-filled bath
- Do not autoclave
Specifica
Specifica
| Descrizione | Bath Armor |
| Tipo | Bath Armor |
| Materiale | Aluminum |
| Quantità | 12 L |
| Forma | Round |