Learn More
VEE GEE Baume Hydrometer, Heavy Liquids
Codice prodotto. p-7250824
Click to view available options
Intervallo Baumé:
0 to 12° subdiv 0.1°
0 to 15° subdiv 0.1°
0 to 25° subdiv 0.2°
0 to 35° subdiv 0.5°
0 to 50° subdiv 0.5°
0 to 50° subdiv 1.0°
0 to 70° subdiv 1.0°
0 to 90° subdiv 1.0°
19 to 31° subdiv 0.1°
29 to 41° subdiv 0.1°
35 to 70° subdiv 0.5°
39 to 51° subdiv 0.1°
9 to 21° subdiv 0.1°
Dimensione della confezione:
Pezzo
Codice prodotto. 17876553
Marca: VEE GEE 66099
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The Baume (Heavy) Hydrometer is for liquids heavier than water.
  • Specific Gravity: 145/(145-degrees Baume)
  • Calibrated for use at 15.5°C/60°F
Specifica
Tipo Baume Hydrometer
Incrementi 0.2°
Lunghezza (sistema metrico) 30.5 cm
Materiale Glass
Intervallo Baumé 0 to 25° subdiv 0.2°
Temperature of Standardization 15.5°C (60°F)