Learn More
BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80
Codice prodotto. p-4869062
Click to view available options
Quantità:
5 lb.
Tipo di imballaggio:
Pail
Dimensione della confezione:
22.30kg
Codice prodotto. 16264202
Marca: BD 212263
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Detection of microorganisms on surfaces sanitized with quaternary ammonium compounds
Certifications
CE, CCAM
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CCAM
Forma Dehydrated
Tipo di media Soy Agar with Lecithin and Polysorbate 80
Product Type Agar
Formulazione Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/mL, Polysorbate 80 5g/L, Agar 15g/L
Da utilizzare con (applicazione) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Tipo di imballaggio Pail
Quantità 5 lb.