Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Chloramphenicol29
Codice prodotto. 11702954
Click to view available options
Concentrazione:
30 μg
Quantità:
10 Pack
Dimensione della confezione:
pezzi 500
Codice prodotto. 11702954
Marca: BD 231274
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifica
Agenti antimicrobici Chloramphenicol29
Concentrazione 30 μg
Tipo di imballaggio Disc
Quantità 10 Pack
For in vitro diagnostic use