Learn More
BCLAF1 Mouse, anti-Human, Unconjugated, Clone: 1D6G7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67860-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
BCLF1, also named as Bcl-2-associated transcription factor 1, is a 920 amino acid protein, which Interacts with Bcl-2 related proteins, EMD, with the adenovirus E1B 19 kDa protein and with DNA. BCLF1 as a death-promoting transcriptional repressor may be involved in cyclin-D1/CCND1 mRNA stability through the SNARP complex which associates with both the 3′end of the CCND1 gene and its mRNA. The calculated molecular weight of BCLF1 is a 110 kDa, but the modified BCLF1 protein is about 140-150 kDa.
Specifica
BCLAF1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
1D6G7 | |
Unconjugated | |
150 μL | |
9774 | |
Liquid |