Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Click to view available options
Quantità:
10 Cartridges (50 Discs/Cartridge)
Dimensione della confezione:
pezzi 10
Descrizione
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifica
Specifica
| Agenti antimicrobici | Ciprofloxacin31 |
| Concentrazione | 5 μg |
| Tipo di imballaggio | Disc |
| Quantità | 10 Cartridges (50 Discs/Cartridge) |
For in vitro diagnostic use