Learn More
BD Cell Viability Solution
Codice prodotto. p-4340669
Click to view available options
Quantità:
100 Tests
500 Tests
Dimensione della confezione:
Pezzo
Codice prodotto. 15878458
Marca: BD 555816
Ready-to-use solution of the nucleic acid dye, 7-Amino-actinomycin D (7-AAD) that can be used in place of propidium iodide (PI) for the exclusion of nonviable cells in flow cytometric assays
  • Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
  • Does not require dilution
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 4°C
Quantità 100 Tests