Learn More
BD Dehydrated Media M17 Agar
Click to view available options
:
500g
Specifica
Specifica
| Content And Storage | <30°C |
| Forma | Dehydrated |
| Product Type | Agar |
| Formulazione | Pancreatic digest of Casein 5g/L, Soy Peptone 5g/L, Beef extract 5g/L, Yeast extract 2.5g/L, Ascorbic Acid 0.5g/L, Magnesium Sulfate 0.25g/L, Disodium-beta-glycerophosphate 19g/L, Agar 11g/L |
| Da utilizzare con (applicazione) | strains used in molecular microbiology procedures |
| Tipo di imballaggio | Bottle |
| Quantità | 500 g |