Learn More
BD Difco™ HYcheck™ Contact Slides: Enterobacteriaceae
Click to view available options
:
Enterobacteriaceae; 10U
Descrizione
- Designed for environmental testing
- Double sided with Tryptic Soy Agar and Violet Red Bile Glucose Agar
- Hinged paddle bends for easy sampling
Order Info
Dieci unità per confezione.
Specifica
Specifica
| Content And Storage | Da 2 ° a 8 °C |
| Forma | Solido |
| Manufacturing Origin | United States |
| Formulazione | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L, Yeast extract 3g/L, Peptone 7g/L, Dextrose 10g/L, Sodium Chloride 5g/L, Bile Salts No. 3 1.5g/L, Neutral red 0.03g/L, Crystal Violet 0.002g/L, Agar 15g/L |
| Da utilizzare con (applicazione) | Assess the microbiological contamination of surfaces or fluids |
| Tipo di imballaggio | Slide |
| Quantità | 10 U |