Learn More
BD PE Mouse anti-Human TGF-β1
Descrizione
The TW4-9E7 monoclonal antibody specifically binds to Human Transforming Growth Factor beta-1 (TGF-β1). TGF-β1 is a potent multifunctional cytokine that positively or negatively regulates numerous processes including development, hematopoiesis, tissue remodeling, wound repair, innate and adaptive immunity as well as cancer and autoimmune diseases. TGF-β1 is formed by the enzymatic cleavage of the TGF-β1 propeptide that is encoded by the TGFβ1 gene and comprised of the Latency Associated Peptide (LAP) and TGF-β1. Prior to secretion, the dimeric LAP-TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β1). This complex may be expressed on the surface of TGF-β1-producing cells or be further processed by proteolytic removal of LAP to release the biologically active mature form of the soluble TGF-β1 homodimer. Many different cell types synthesize TGF-β1 and express specific receptors for it.
- Alternate name: TGFB1; TGFβ1; TGF-β-1; Transforming growth factor, β 1; CED; LAP
- Clone: TW4-9E7
- Immunogen: Human TGF-β1 transfected cell line
- Isotype: Mouse (BALB/c) IgG1, κ
- Strain host: BALB/c
- Reactivity: Human
- Format: PE
- Volume: 5μL
- Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Intracellular staining (flow cytometry), Flow cytometry
Specifica
Specifica
| Da utilizzare con (applicazione) | Flow Cytometry, Intracellular Staining |
| Formulazione | Aqueous Buffered solution containing BSA and 0.09% Sodium Azide |
| Peso molecolare | 14.5 |
| Nome | Mouse anti-Human TGF-beta1 |
| Quantità | 100 Tests |
| Sorgente | BALB/c |
| Requisiti di stoccaggio | 4°C |
| Coniugato | PE Labeled |
| Cross-reattività | Mouse |
| Forma | Liquid |
Research use only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.