Learn More
BD Prepared Bottled Media: Trypticase™ Soy Broth (Double Wrapped) Infusion Bottle
Click to view available options
:
Trypticase Soy Broth; 125mL
Descrizione
Ready for immediate use
Specifica
Specifica
| Colore | Amber |
| Content And Storage | 5°C to 25°C |
| Forma | Liquid |
| Manufacturing Origin | United States |
| Tipo di media | Trypticase Soy Broth |
| Volume di riempimento | 100 mL |
| Formulazione | Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Glucose (Dextrose) 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L |
| Da utilizzare con (applicazione) | Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples |
| Tipo di imballaggio | Bottle |
| Quantità | 10/Pk. |