Learn More
behr Labor-Technik™ behrotest™ Reflux Condenser Allhin
Prevent solvents from boiling away. behr Labor-Technik™ behrotest&tradeReflux Condenser Allhin is used to prevent solvent from boiling away. As vapor from the boiling solvent rises into inside column of the reflux condenser, it is cooled by the jacket of water on the outside and condenses.
Marca:  behr Labor-Technik™ B00217957
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 10004771
  • 216.00€ / Pezzo
Spedizione stimata: 30-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • With oval bulbs
Specifica
Specifica
2.7 L
0.48 m
0.045 m
Width: 0.125m, Height: 0.045m, Depth: 0.48m
0.125 m