Learn More
Bel Art™ Rack per colture di tessuti Stak-a-Tray™ SP Scienceware™
Sfoglia Tutto SP Scienceware Prodotti
Click to view available options
Product Type:
Large Tray Holder
Small Tray Holder
Vassoio, intero
Capacità:
Microplates (Standard Size Flat)
Sample Containers
Dimensione della confezione:
Pezzo
Descrizione
- Keeps trays, flasks and dishes level and stable for even settling of tissue culture cells
- Racks interlock when stacked, to a maximum of 5 racks high (allows up to 20 levels of trays)
- Each rack tier holds one large tray (Mfr. No. 18610-1440), 35.6 x 35.6cm, or two smaller trays (Mfr. No. 18610-0470), 17.8 x 35.6cm
- Perforated trays allow air, gas and water vapor circulation
- Racks and trays sold separately
- Stainless-steel storage system is autoclavable
Rack, 2-Tier (Mfr. No. 18610-0420)
- 5cm clearance between trays, ideal for storing larger culture flasks
Rack, 4-Tier (Mfr. No. 18610-0440)
- 2.4cm clearance between trays, for storing Petri dishes or smaller culture flasks
Specifica
Specifica
| Product Type | Small Tray Holder |
| Matrice | 12 x 1 |
| Autoclavabile | Autoclavable |
| Colore | Silver |
| Capacità | Sample Containers |
| Materiale | Stainless Steel |
| Posizioni | 12 |
| Forma | Rettangolare |
| Ripiani | None |
| Sovrapponibile | Sì |
| Vedi altri risultati |