Learn More
Cytiva Benzamidine Sepharose™ 4 Fast Flow, High Sub Media
Sfoglia Tutto Cytiva Prodotti
Click to view available options
:
25mL
Descrizione
- p-aminobenzamide (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity and allows easy scale-up
- Binding capacity ≥35mg trypsin/mL media
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
Specifica
Specifica
| Dimensione particelle | 90 μm |
| pH | 2 to 8 (long term); 1 to 9 (short term) |
| Descrizione | 25 mL |
| Materiale | Benzamine Sepharose 4 Fast Flow |