Learn More
beta 2-Microglobulin Antibody (B2M-01) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB500-317
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Bovine (Negative), Canine (Negative), Chicken (Negative), Mouse (Negative), Rabbit (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Radioimmunoassay.
Specifica
| Monoclonal | |
| 12 kDa | |
| Store at 4C. Do not freeze. | |
| IgG2a |
| B2M | |
| Protein A purified | |
| Purified |