missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta galactosidase Mouse anti-Human, Clone: 4F4F4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66586-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GLB1 cleaves beta-linked terminal galactosyl residues from gangliosides, glycoproteins, and glycosaminoglycans. Isoform 2 has no beta-galactosidase catalytic activity, but plays functional roles in the formation of extracellular elastic fibers (elastogenesis) and in the development of connective tissue. This protein seems to be identical to the elastin-binding protein (EBP), a major component of the non-integrin cell surface receptor expressed on fibroblasts, smooth muscle cells, chondroblasts, leukocytes, and certain cancer cell types. In elastin producing cells, associates with tropoelastin intracellularly and functions as a recycling molecular chaperone which facilitates the secretions of tropoelastin and its assembly into elastic fibers.Specifica
| Beta galactosidase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P16278 | |
| GLB1 | |
| Beta galactosidase Fusion Protein Ag8069 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4F4F4 | |
| Unconjugated | |
| GLB1 | |
| Acid beta galactosidase, Beta galactosidase, EBP, Elastin receptor 1, ELNR1, galactosidase, beta 1, GLB1, Lactase | |
| Mouse | |
| Protein G | |
| RUO | |
| 2720 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Ser du en mulighed for forbedring?Del en indholdskorrektion