Learn More
E in., celon Bev-A-Line™ IV Tubing
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1/2 in.
1/4 in.
1/8 in.
3/4 in.
3/8 in.
Diametro (sistema imperiale) esterno:
1 in.
1-1/4 in.
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/8 in.
Dimensione della confezione:
15m
Descrizione
Meets FDA, USDA, National Formulary, and USP standards for food, beverage, pharmaceutical, and laboratory usage. Co-e in., truded to combine the benefits of two tubes in one product. The clear shell provides e in., cellent durability and greater burst strength than PVC. Complete visual flow. Free technical application support available to assist with tubing selection
Specifica
Specifica
| Diametro (sistema imperiale) interno | 1/8 in. |
| Diametro (sistema imperiale) esterno | 3/16 in. |
| Overall Length (English) | 50 ft. |
| Colore | Natural |
| Product Type | Bev-A-Line IV Tubing |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.