Learn More
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Dimensioni (L x P):
8 x 8 in.
9 x 9 in.
12 x 12 in.
15 x 15 in.
18 x 18 in.
20 x 20 in.
24 x 24 in.
30 x 30 in.
36 x 36 in.
40 x 40 in.
45 x 45 in.
45 x 48 in.
54 x 54 in.
54 x 72 in.
Dimensione della confezione:
pezzi 100
pezzi 1000
pezzi 250
Descrizione
- Designed for steam sterilization
- Treated to impart water and alcohol repellency
- Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
- Sufficient microbial barrier
- Resists tearing and punctures
- Does not contain any natural latex rubber
- Processed and packaged in ISO Certified Cleanrooms
- Certificate of Conformance included with shipment
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Certificazione/Conformità | COC |
| Tipo di chiusura | None |
| Colore | Blue |
| Monouso | Single-use |
| Da utilizzare con (applicazione) | Steam Sterilization |
| Da utilizzare con (apparecchiatura) | Pharmaceutical, Hospital, Medical, Dental |
| Materiale | Cellulose |
| Sterilità | Non-sterile |
| Senza lattice | Yes |
| Vedi altri risultati |