Learn More
Tocris Bioscience™ BI 2852 FL
Click to view available options
:
100 μg
Descrizione
BI 2852 FL is a cell-permeable pan-RAS bioluminescence resonance energy transfer (BRET) fluorescent probe. BI 2852 FL is synthesized by conjugating a fluorophore to a derivative of the reversible SI/II-P KRAS inhibitor, BI 2852. BI 2852 FL is used in competitive BRET format to quantify SII-P engagement to multimeric RAS complexes in live cells.
Specifica
Specifica
| Quantità | 100 μg |
| Content And Storage | Store at -20°C |
| Bersaglio | Other Fluorescent Probes |