Learn More
Zeiss™ Binocular Phototube
Click to view available options
Tipo:
Binocular Phototube 20°/23 (100:0/0:100)
Binocular Phototube 30°/23 (100:0/0:100)
Binocular Phototube 30°/23 (50:50)
Binocular Phototube S 20°
Binocular Phototube, Left Port 45°/23 (50:50)
Binocular Tube 20°/23
Binocular Tube 30°/20
Dimensione della confezione:
Pezzo
Descrizione
Binocular phototubes are available with a variety of options to suit your needs
Specifica
Specifica
| Tipo | Binocular Phototube 30°/23 (100:0/0:100) |
| Descrizione | Reversed image with sliding prism, low-vibration prism switch; Camera port with interface 60N |
| Quantità | 1 Ea. |