Learn More
Ansell BioClean Isolator Sleeve - Glove Assembly Unit C Ring and O Ring BBCO 100
Click to view available options
Da utilizzare con (apparecchiatura):
Gloves, Sleeves
Dimensione della confezione:
Pezzo
Descrizione
- Enhanced convenience and reliability: Made of PVC and silicone, the BioClean™ Isolator Sleeve/Glove Assembly is specially designed to enable the easy connection of gloves to isolator sleeves
- Improved hygiene: This isolator glove assembly unit is double-bagged in EasyTear packaging, ensuring hygiene and convenience
Specifica
Specifica
| Da utilizzare con (applicazione) | Connecting glove to isolator sleeve |
| Da utilizzare con (apparecchiatura) | Gloves, Sleeves |
| Product Type | O-Ring and C-Ring Set |
| Materiale | Silicone, PVC |
| Size | 11 |