Learn More
Corning™ Biocoat™ collagen V, human, 0.25mg
Sfoglia Tutto Corning Prodotti
Click to view available options
Quantità:
0.25 mg
Dimensione della confezione:
250µg
Descrizione
Corning Collagen V, human, is used as a thin coating on tissue-culture surfaces to study collagen V effects on cell behaviour.
- Sourced from human placenta. NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody
- Electrophoretically homogeneous (SDS-PAGE)
- Tested and found negative for bacteria, fungi, and mycoplasma
- Stable for at least three months at -20°C. Avoid multiple freeze-thaws
- Recommended concentration - 1-5 μg/cm2 of growth surface depending on cell type
- Formulation - frozen in 10mM acetic acid
Specifica
Specifica
| Quantità | 0.25 mg |