Learn More
Corning™ Biocoat™ tumour necrosis factor-a (TNF-a), human recombinant, 50μg
Sfoglia Tutto Corning Prodotti
Click to view available options
Quantità:
5 x 10 μg
Dimensione della confezione:
50µg
Descrizione
Corning Tumour Necrosis Factor-a (TNF-a) has growth-inhibitory effects on many tumour cells and nontumour cells such as certain haematopoietic cells. The product also has differentiation-inducing effect, on macrophages.
Specifica
Specifica
| Quantità | 5 x 10 μg |