Learn More
Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters
Codice prodotto. p-4527816 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection Tube
100 mL Round-bottom, Open-tube
15 mL Conical Tube
15 mL Starstedt Tube
150 mL Round-bottom, Open-tube
180 mL Polypropylene Screw Cap Bottle
25 mL DIN Round-bottom Tube
25 mL Universal Tube
5/7 mL Blood Collection Tube
5/7 mL Round-bottom, Open-tube
50 mL Conical Tube
50 mL DIN Round-bottom Tube
Dimensione della confezione:
pezzi 4
Codice prodotto. 10247154
Marca: Thermo Scientific™ 75003677
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
TRUSTED_SUSTAINABILITY
Specifica
Tipo Rotor Tube Adapter
Tipo di provetta Conical
Capacità 50 mL Conical Tube
Dimensioni provetta 50 mL