Learn More
ACROBiosystems Biotinylated Human Angiopoietin-like 3 / ANGPTL3 Protein, His, Avitag™
Biotinylated Protein
457.25€ - 2011.89€
Specifica
Numero di accesso | NP_055310 |
---|---|
Peso molecolare | 27.4 kDa |
Sorgente | Biotinylated Human Angiopoietin-like 3, His,Avitag (AN3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession # Q9Y5C1-1). |
Immunogeno | Ser 17 - Pro 220 |
Requisiti di stoccaggio | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17116071
|
ACROBiosystems
AN3-H82E7-200UG |
200 μg |
2011.89€
200µg |
Spedizione stimata: Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17126071
|
ACROBiosystems
AN3-H82E7-25UG |
25 μg |
457.25€
25µg |
Spedizione stimata: Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Angiopoietin-like protein 3 (ANGPTL3) is also known as Angiopoietin-related protein 3, Angiopoietin-5 (ANGPT5 / ANG-5), is a member of the angiopoietin-like family of secreted factors. ANGPTL3 / ANGPT5 is predominantly expressed in the liver, and has the characteristic structure of angiopoietins, consisting of a signal peptide, N-terminal coiled-coil domain and the C-terminal fibrinogen (FBN)-like domain. The FBN-like domain in angiopoietin-like 3 protein was shown to bind alpha-5/beta-3 integrins, and this binding induced endothelial cell adhesion and migration. This protein may also play a role in the regulation of angiogenesis. Angptl3 also acts as dual inhibitor of lipoprotein lipase (LPL) and endothelial lipase (EL), and increases plasma triglyceride and HDL cholesterol in rodents. ANGPTL3 inhibit endothelial lipase to catalyze HDL-phospholipid and increase HDL-PL levels. Circulating PL-riched HDL particles have high cholesterol efflux abilities.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.4 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_055310 | |
Biotinylated Human Angiopoietin-like 3, His,Avitag (AN3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession # Q9Y5C1-1). | |
-20°C | |
Angiopoietin-like 3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
27.4 kDa | |
Ser 17 - Pro 220 | |
ANGPTL3,ANGPT5,ANG-5,Angiopoietin-5,FHBL2 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |