Learn More
ACROBiosystems Biotinylated Human IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, Fc, Avitag&Fc, Avitag (MALS verified)
Marca: ACROBiosystems ILG-H82F4-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.
Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 52.8 kDa (IL-2RB) and 77.1 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terSpecifica
NP_000869.1 (IL-2 RB) & NP_000408.1 (IL-2 RA) & AAH14972.1 (IL-2 RG) | |
200 μg | |
Ala 27 - Asp 239 & Glu 22 - Ser 212 & Leu 23 - Asn 254 | |
IL-2 R beta & IL-2 R alpha & IL-2 R gamma | |
Precision Avi | |
Fc Tag & Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE. |
52.8 kDa/77.1 kDa | |
Biotinylated Human IL-2 RB&IL-2; RA&IL-2; RG, Fc,Avitag&Fc;,Avitag (ILG-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 & Glu 22 - Ser 212 & Leu 23 - Asn 254 (Accession # P14784-1 (IL-2 RB) & P01589-1 (IL-2 RA) & P31785-1 (IL-2 RG)). | |
-20°C | |
IL-2 R beta & IL-2 R alpha & IL-2 R gamma | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |