Learn More
ACROBiosystems Biotinylated Human IL-8 protein, His, Avitag
Marca:  ACROBiosystems IL8-H82H3-25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17286260
  • 457.25€ / 25µg
Spedizione stimata: 07-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Interleukin 8 (IL8 or chemokine (C-X-C motif) ligand 8, CXCL8) is a chemokine produced by macrophages and other cell types such as epithelial cells, airway smooth muscle cells and endothelial cells. There are many receptors on the surface membrane capable of binding IL-8, the most frequently studied types are the G protein-coupled serpentine receptors CXCR1 and CXCR2. Expression and affinity for IL-8 differs between the two receptors (CXCR1 > CXCR2). IL-8(6-77) has a 5-10-fold higher activity on neutrophil activation, IL-8(5-77) has increased activity on neutrophil activation and IL-8(7-77) has a higher affinity to receptors CXCR1 and CXCR2 as compared to IL-8(1-77), respectively.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 10.3 kDa.
Specifica
Specifica
NP_000575.1
25 μg
Ser 28 - Ser 99
CXCL8,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1
Precision Avi
His Tag & Avi Tag
Powder
10.3 kDa
Biotinylated Human IL-8, His,Avitag (IL8-H82H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).
-20°C
IL-8
Human
HEK293
MABSol™ Biotin Labeled Proteins