Learn More
Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Dimensione chiusura:
10 mm
15 mm
18 mm
22 mm
28 mm
Dimensione della confezione:
pezzi 300
Descrizione
- Specifically designed for use with Fisherbrand™ Glass Screw Neck Specimen Vials
- Polypropylene construction with foil lining
- Fit both tall and squat form vials
- Available in a variety of sizes to match vial specifications
- Screw neck specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
Specifica
Specifica
| Colore | Nero |
| Materiale | Polipropilene con rivestimento in AFM |
| Tipo di chiusura | Tappo a vite |
| Dimensione chiusura | 22 mm |